Learn More
Description
- Produces a pink appearance in nuclei
- Suitable for immunohistochemistry, immunocytochemistry/immunofluorescence, and in situ hybridization
- Ready-to-use formulation
- Excellent contrast in multiple labeling applications when hematoxylin obscures substrate colors
- Rapid, single-step staining procedure
- Compatible with non-aqueous mountants
Specifications
Specifications
| For Use With (Application) | Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, In-Situ Hybridization |
| Product Line | Novus |
| Quantity | 500 mL |
| Storage Requirements | Room temperature |
| Product Type | Counterstain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
