Learn More
United Scientific Supplies Nuclear Magnetic Resonance Apparatus
SDPSCI_ED
Catalog No. S07395 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Nuclear magnetic resonance apparatus
Catalog No.
S07395 Nuclear magnetic resonance apparatus
1 options
Catalog No. S07395 Supplier United Scientific Supplies Supplier No. NMRA01
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Permanent magnet, oscillator, frequency counter, and power supply
Requires
Oscilloscope
Set of equipment for studying the nuclear magnetic resonance of protons in hydrogen.
  • Allows students to measure the gyromagnetic ratio, Lande factor, and nuclear magnetic moment
Specifications
Product Type Nuclear Magnetic Resonance Apparatus
Includes Permanent magnet, oscillator, frequency counter, and power supply