Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nuclear Matrix Protein p84 Monoclonal Antibody (5E10), HRP
GREENER_CHOICE

Catalog No. PIMA531590 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531590 Supplier Invitrogen™ Supplier No. MA531590
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Positive Control: Jurkat, Raji, NCI-H929, HeLa, HeLa nuclear. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Matrix Protein p84
Applications Western Blot
Classification Monoclonal
Clone 5E10
Concentration 0.3 mg/mL
Conjugate HRP
Formulation TBS with 1% BSA and 0.005% thimerosal; pH 7.4
Gene Thoc1
Gene Accession No. Q96FV9
Gene Alias 3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1
Gene Symbols Thoc1
Host Species Mouse
Immunogen Amino acids 15-374 of human p84 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.