Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nuclear Matrix Protein p84 Monoclonal Antibody (5E10)
GREENER_CHOICE

Catalog No. PIMA123261 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA123261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA123261 Supplier Invitrogen™ Supplier No. MA123261
  • $715.00
    $557.65 / Each of 1

    Save $157.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Suggested positive controls are HeLa, Raji, and MOLT4.

HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Matrix Protein p84
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7
Gene Thoc1
Gene Accession No. P59924, Q8R3N6, Q96FV9
Gene Alias 3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1
Gene Symbols Thoc1
Host Species Mouse
Immunogen Amino acids 15-374 of human p84 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100770844, 225160, 291797, 9984
Target Species Human, Mouse, Rat, Monkey, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.