Learn More
Invitrogen™ Nuclear Receptor Buffer K
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Specifications
Specifications
| Physical Form | Liquid |
| Product Line | LanthaScreen |
| Quantity | 25 mL |
| Grade | Research Grade |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.