Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclease-Free Water, Promega™

Catalog No. prp1197 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
150 mL
2 x 25 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRP1197 500 mL
PR-P1193 2 x 25 mL
PRP1195 150 mL
PRP1199 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRP1197 Supplier Promega Corporation Supplier No. P1197
Only null left
Add to Cart
Add to Cart

Quality tested and certified free of DNase and RNase activity.

  • Quality tested and certified free of DNase and RNase activity

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Quantity 500 mL
Chemical Name or Material Water, Nuclease free
Recommended Storage Below 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.