Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Nuclease P1, MP Biomedicals™

Catalog No. MP218055501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218055501 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP218055501 Supplier MP Biomedicals™ Supplier No. 0218055501
Add to Cart
Add to Cart

Exhibits both phosphodiesterase and monoesterase activities. Nuclease P1, MP Biomedicals is a single strand-specific nuclease that exhibits high selectivity for single-stranded nucleic acids and single-stranded regions in double-stranded nucleic acids. De

Catalyzes the nonspecific endonucleolytic cleavage of single stranded DNA and RNA to yield nucleoside 5′-phosphates and 5′-phosphooligonucleotides. It does not appreciably degrade double-stranded nucleic acids, especially in the presence of more than 400 mM sodium chloride at pH 6.0.

  • Useful at removing single stranded strands hanging off the end of double stranded DNA and at completely cleaving melted DNA for simple DNA composition analysis
  • Improves the sensitivity of a 32P-labeling method for the detection of DNA adducts
  • Used in a study to assess crystal structures using ammonium sulphate or polyethylene glycol 4000 as a precipitating agent
  • Used in a study to investigate a method for the direct sequence analysis 20-25 nucleotides from the terinini of 5′ or 3′ end group labeled RNA
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH Optimum pH: 5.3 (RNA and heat-denatured DNA) and 7.2 (3′-AMP) (Lit.)
Content And Storage 4°C
Quantity 1 Vial
Form Solid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.