Learn More
ACROBiosystems Nucleocapsid protein 200ug
Supplier:  ACROBiosystems NUNC81Q6200UG
Encompass_Preferred
Biotinylated SARS-CoV-2 Nucleocapsid protein HisAvitag (NUN-C81Q6) is expressed from E. coli cells. It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).
Catalog No. 50-253-4354
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.