Learn More
Description
Recommended positive controls: Raji cell extract. This antibody does not react with mouse protein.
Specifications
Specifications
| Antigen | Nucleolin |
| Applications | ChIP Assay, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4E2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and no preservative |
| Gene | NCL |
| Gene Accession No. | P19338 |
| Gene Alias | B530004O11Rik; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; Ncl; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
