Learn More
Description
Includes
Reagents A and B, 5.0M Sodium Perchlorate, and Nucleon resins.
- Phenol-free protocol requiring only 30 minutes to complete
- Low-shearing protocol gives maximum recoveries of high-molecular weight DNA
- For 50 preps of 10mL of whole blood (sufficient quantities of all reagents provided)
- Available in different formats/reaction scales (see 45-000-955 and 45-000-956)
Specifications
Specifications
| Description | illustra Nucleon BACC system rapidly extracts high molecular weight DNA from whole blood and cultured cells. |
| Format | Kit |
| Isolation Technology | Organic Extraction |
| Sample Type | Blood, Cell Cultures |
| For Use With (Application) | Rapid extraction of high-quality, high molecular weight genomic DNA from cell cultures and blood. |
| Includes | Reagents A and B, 5M Sodium Perchlorate and Nucleon™ Resins |
| Product Type | illustra Nucleon BACC3 |
| Quantity | 50 Kits |
| Yield | 370-440 μg DNA/mg tissue |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.