Learn More
Macherey-Nagel HPLC column EC 250/4.6 NUCLEOSIL 120-5 C18
Supplier:  Macherey-Nagel 720041.46
Encompass_Preferred
EC HPLC column - EC 250/4.6 NUCLEOSIL 120-5 C18 - length: 250 mm, ID: 4.6 mm - pack of 1
Catalog No. NC0223297
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.