Learn More
Macherey-Nagel EC HPLC column (analytical), NUCLEOSIL 100-3 C18, 3 um, 250x4.6 mm
Supplier:  Macherey-Nagel 720133.46
Encompass_Preferred
This HPLC column is packed with endcapped C18 (ODS, octadecyl silyl) modification, based on the original NUCLEOSIL silica gel. It is one of the first spherical silicas for HPLC.
Catalog No. NC1887203
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.