Learn More
Macherey-Nagel NucleoSpin 8 RNA (12x8)
Supplier:  Macherey-Nagel 740698
Encompass_Preferred
NucleoSpin 8 RNA (12 x 8) 12 x 8 preps for the isolation of RNA from cells and tissue - NucleoSpin RNA Binding Strips, MN Wash Plates, MN Square-well Blocks , Racks of Tube Strips, Elution Plate U-bottom, Self-adhering Foil, buffers, RNase-free rDNase
Catalog No. NC1438368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.