Learn More
Sartorius NUCLIGHT LENTIVIRUS 2.0 EFLA
Supplier:  Sartorius BA04888
Encompass
Incucyte® Nuclight Lentivirus 2.0 (EF1a, Puro)One vial of lentivirus reagent (>1x106 TU/mL) suitable for transducing cells with a non-perturbing, nuclear restricted fluorescent label driven off an EF1α promoter. Stable cell lines can be created under puromycin selection.
Catalog No. NC3871994