Learn More
Sartorius Incucyte Nuclight Red Lentivirus 2.0, Efficient Nuclear Labeling, Minimal Toxicity, 0.6 mL
Catalog No. NC3706347 Shop All Sartorius Products
Encompass
The Incucyte Nuclight Red Lentivirus 2.0 is an innovative reagent designed for efficient nuclear labeling of live mammalian cells. By utilizing a third-generation, HIV-based lentivirus system, it enables real-time visualization and quantification of cell proliferation and viability with minimal toxicity. Ideal for generating stable cell populations with puromycin selection, it is fully compatible with Incucyte Live-Cell Analysis Systems, providing flexibility for various assays in live-cell studies.
Catalog No. NC3706347 Supplier Sartorius Supplier No. BA04887
The Incucyte Nuclight Red Lentivirus 2.0 is an innovative reagent designed for efficient nuclear labeling of live mammalian cells. By utilizing a third-generation, HIV-based lentivirus system, it enables real-time visualization and quantification of cell proliferation and viability with minimal toxicity. Ideal for generating stable cell populations with puromycin selection, it is fully compatible with Incucyte Live-Cell Analysis Systems, providing flexibility for various assays in live-cell studies.