Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDT13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559234 Supplier Invitrogen™ Supplier No. PA559234
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDAQRIEDSV LIGCSEQQEA WFALDLGLDS SFSISASLHK PEMETELKGS FIELRKALFQ LNARDASLLS TAQALLRWH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 73%.

NUDT13 (Nudix Hydrolase 13) is a Protein Coding gene. Among its related pathways areSynthesis and interconversion of nucleotide di- and triphosphatesandPurine metabolism (REACTOME). Gene Ontology (GO) annotations related to this gene includehydrolase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT13
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUDT13
Gene Accession No. Q86X67
Gene Alias 4933433B15Rik; LOW QUALITY PROTEIN: nucleoside diphosphate-linked moiety X motif 13; nucleoside diphosphate linked moiety X motif 13; nucleoside diphosphate-linked moiety X motif 13; Nucleoside diphosphate-linked moiety X motif 13-like protein; nudix (nucleoside diphosphate linked moiety X)-type motif 13; nudix hydrolase 13; nudix motif 13; nudix-type motif 13; NUDT13; Protein KiSS-16; Unknown (protein for MGC:140322)
Gene Symbols NUDT13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUDT13. Recombinant protein control fragment (Product #RP-97164).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25961
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.