Learn More
Enzo Life Sciences Nullscript (1mg). CAS: 300816-11-9
SDP
Supplier:  Enzo Life Sciences BMLGR3270001
Encompass_Preferred
Negative control compound for the histone deacetylase (HDAC) inhibitor scriptaid (BML-GR326). Alternative name: 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide. Purity: ≥97% (TLC). Solubility: Soluble in DMSO (4mg/ml) or ethanol (0.3mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0982
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.