Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Nullscript, Inactive HDAC Inhibitor, 298.3 g/mol, 5 mg

Catalog No. 502706486
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6486 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6486 Supplier Abcam Supplier No. AB1416395MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inactive analog of histone deacetylase inhibitor scriptaid used as a negative control.

  • Molecular Weight: 298.3 g/mol
  • Purity: >97%
  • Description: Inactive analog of histone deacetylase (HDAC) inhibitor scriptaid (ab120883), used as a negative control.
  • Storage: Store at -20°C.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Histone Deacetylase
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Formulation 100% Nullscript
Quantity 5 mg
For Use With (Application) Negative control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.