Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NuMA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587448 Supplier Invitrogen™ Supplier No. PA587448
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTLHATRGAA LLSWVNSLHV ADPVEAVLQL QDCSIFIKII DRIHGTEEGQ QILKQPVSER LDFVCSFLQK NRKHPSSPEC LVSAQKVLEG SELELAKMTM LLLYHSTMSS KSPRDWEQFE YKIQAELAVI LKFVLDHEDG LNLNEDLENF LQKAPVPSTC SSTFPEELSP PSHQAKREIR FLELQKVASS SSGNNFLSGS PASPMGDILQ TPQFQMRRLK KQLADERSNR DELELELAEN RKLLTEKDAQ IAMMQQRIDR LALLNEKQAA SPLEPKELEE LRDKNESLTM RLHETLKQCQ DLKTEKSQM; Positive Samples: HeLa, Mouse kidney; Cellular Location: Chromosome, Cytoplasm, Nucleus matrix, cytoskeleton, spindle pole.

NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NuMA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NUMA1
Gene Accession No. E9Q7G0, Q14980
Gene Alias 6720401E04Rik; AA764025; AL022610; AU014979; centrophilin stabilizes mitotic spindle in mitotic cells; NMP22; NMP-22; Nuclear matrix protein-22; Nuclear mitotic apparatus protein; Nuclear mitotic apparatus protein 1; NUMA; NuMA protein; Numa1; SP-H antigen; structural nuclear protein
Gene Symbols NUMA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-309 of human NUMA1 (NP_0061762).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101706, 308870, 4926
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.