Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ 2 mL Internally-Threaded Universal Tubes
GREENER_CHOICE

Catalog No. 12590011
Change view
Click to view available options
Barcode:
2D Barcode
2D Barcode (Rack Only)
2D Barcode (Tube Only)
None
Product Type:
Universal Tube
Universal Tubes in Latch Rack
Packaging:
10 Racks of 48 Tubes
Bulk
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Barcode Product Type Packaging
12-590-011 None Universal Tubes in Latch Rack 10 Racks of 48 Tubes
12-590-012 2D Barcode Universal Tube Bulk
12-590-013 None Universal Tube Bulk
12-590-010 2D Barcode (Tube Only) Universal Tubes in Latch Rack 10 Racks of 48 Tubes
12-565-998 2D Barcode Universal Tubes in Latch Rack 10 Racks of 48 Tubes
12-565-999 2D Barcode (Rack Only) Universal Tubes in Latch Rack 10 Racks of 48 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12-590-011 Supplier Thermo Scientific™ Supplier No. 374511
Add to Cart
Add to Cart

Experience the best features of our storage tube portfolio: universal cap, universal latch rack and free-standing tube design -- all for either benchtop or automation use.

Experience the best features of the Thermo Scientific™ storage tube portfolio: universal cap, universal latch rack and free-standing tube. This versatile, 2.0mL internally threaded tube is ideal both for a large storage system like a biobank, and in smaller scale for traditional cryostorage. Sample identity is ensured due to a permanently attached 2D barcode on each tube and permanent laser etching of the rack. Now available in a barcoded rack to allow tracking of the rack in addition to individual tubes.

Scalable

  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10x10 arrays
  • Internal threads limit contamination
  • Automation-pickable in universal latch rack
  • Barcoded rack option offers additional tracking assurance

Innovative Universal Cap Design

  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen

Secure Tracking

  • Permanently bonded, unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 10-6 and is made with class VI medical grade resins
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Closure Color Natural
Capacity (Metric) 2.0 mL
Bottom Shape Flat
Material Plastic
Thread Style Internal
For Use With (Equipment) Biobank, Cryostorage
Packaging 10 Racks of 48 Tubes
Barcode None
Product Line Nunc
Sterility Sterile SAL 10-6
Graduated Yes
Product Type Universal Tubes in Latch Rack
Marking Spot Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.