Learn More
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate
Catalog No. 12565295
Change view
Click to view available options
No. of Wells:
96
384
No. per Pack:
1 Microplates
10 Microplates
25 Microplates
No. per Case:
100 Microplates
30 Microplates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells No. per Pack No. per Case
12-565-295 96 1 Microplates 30 Microplates
12-565-294 384 10 Microplates 30 Microplates
12-567-150 384 25 Microplates 100 Microplates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-565-295 Supplier Thermo Scientific™ Supplier No. 242765
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems.
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.
  • Clear Polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Customized bar coding is available
  • Flexible working volumes; 10-100 μL
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format SBS Standard
Lid With Lid
Culture Area 0.084 cm2/well
Description Non-Treated; w/ lid; Sterile; 1/30
Array 16 x 24
Packaging 1/Pk., 30/Cs.
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 120 μL
No. of Wells 96
Volume (Metric) Working 10 to 100 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 30 Microplates
No. per Pack 1 Microplates
Quantity 1 Ea.
Show More Show Less
Should I use an opaque black/white immuno plate, or an "optical-bottom" design?
Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.