Learn More
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Catalog No. 12565373 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Blue
Natural
Red
White
Yellow
Sterility:
Non-sterile
Sterile
Well Shape:
Round
Round, Conical Bottom (V-well)
Fit for Purpose:
Certified Quality
Sterile
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Well Shape Fit for Purpose
12-565-373 Blue Non-sterile Round Certified Quality
12-565-435 Red Non-sterile Round, Conical Bottom (V-well) Certified Quality
12-565-436 Natural Non-sterile Round, Conical Bottom (V-well) Certified Quality
12-565-437 Black Non-sterile Round, Conical Bottom (V-well) Certified Quality
12-565-438 Natural Sterile Round, Conical Bottom (V-well) Sterile
12-565-439 Yellow Non-sterile Round, Conical Bottom (V-well) Certified Quality
12-565-440 White Non-sterile Round, Conical Bottom (V-well) Certified Quality
12-565-441 Blue Non-sterile Round, Conical Bottom (V-well) Certified Quality
12-565-368 Natural Non-sterile Round Certified Quality
12-565-369 Natural Sterile Round Certified Quality
12-565-370 Black Non-sterile Round Certified Quality
12-565-371 White Non-sterile Round Certified Quality
12-565-263 Natural Non-sterile Round, Conical Bottom (V-well) Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 12-565-373 Supplier Thermo Scientific™ Supplier No. 267385
Add to Cart
Edge
Add to Cart
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
  • Shared-wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance Certified RNase and DNase Free. ANSI Compliant.
Format Standard
Lid Without Lid
Description 96-well Polypropylene Sample Processing & Storage Plates
Array 8 x 12
Autoclavable Autoclavable
Packaging 10 pk., 120 cs.
Binding Type Low binding PP
Product Line Nunc
Color Blue
Product Type 96-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 500 μL
No. of Wells 96
Volume (Metric) Working 450 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Round
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
No. per Case 120
No. per Pack 10
Quantity 120/Cs.
Regulatory Status RUO
RNase Free Yes
Fit for Purpose Certified Quality
Plate Size ANSI/ SLAS Standard