Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cryo Vial Closure Color Coders
GSA_VA

Catalog No. 12565246
Change view
Click to view available options
Color:
Assorted
Blue
Brown
Gray
Green
Orange
Pink
Purple
Red
White
Yellow
Product Line:
Nalgene
Nunc
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Line
12-565-246 Yellow Nunc
12-565-242 White Nunc
12-565-243 Blue Nunc
12-565-244 Red Nunc
12-565-245 Green Nunc
12-565-247 Orange Nunc
12-565-251 Brown Nunc
12-565-252 Pink Nunc
12-565-253 Gray Nunc
12-565-254 Purple Nunc
12-565-180 Assorted Nunc
15-350-47 Blue Nalgene
15-350-45 White Nalgene
15-350-46 Yellow Nalgene
15-350-48 Green Nalgene
15-350-49 Red Nalgene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 12-565-246 Supplier Thermo Scientific™ Supplier No. 355077
Add to Cart
Add to Cart

Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.

Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.

  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Nunc™ CryoColor Vial Closure Coder, PS
Color Yellow
Material Polystyrene
Product Line Nunc
Quantity 500/Pk., 2000/Cs.
For Use With (Equipment) Cryogenic Vial
Product Type Vial Closure
Are your Nunc CryoColor Vial Closure Coders removable?

Once inserted, the Nunc CryoColor Vial Closure Coders cannot be removed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.