Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes
GREENER_CHOICE

Catalog No. 12565299
Change view
Click to view available options
Bottom Shape:
Conical (Self-standing)
Round
Round (Self-standing)
Capacity (Metric):
0.5 mL
1 mL
1.8 mL
3.6 mL
4.5 mL
Thread Style:
External
Internal
Style:
No Writing Surface, A-R Base
Writing Surface
Writing Surface, A-R Base
Writing Surface, No Cap
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottom Shape Capacity (Metric) Thread Style Style
12-565-299 Round (Self-standing) 4.5 mL External No Writing Surface, A-R Base
12-570-216 Conical (Self-standing) 0.5 mL Internal No Writing Surface, A-R Base
12-570-217 Conical (Self-standing) 1 mL Internal Writing Surface
12-565-164N Conical (Self-standing) 1 mL External Writing Surface, A-R Base
12-565-297 Conical (Self-standing) 1 mL External No Writing Surface, A-R Base
12-565-168N Conical (Self-standing) 1 mL Internal Writing Surface, A-R Base
12-565-169N Conical (Self-standing) 1 mL Internal Writing Surface
12-565-163N Round (Self-standing) 1.8 mL External Writing Surface, A-R Base
12-565-298 Round (Self-standing) 1.8 mL External No Writing Surface, A-R Base
12-565-167N Round (Self-standing) 1.8 mL Internal Writing Surface, A-R Base
12-565-171N Round (Self-standing) 1.8 mL Internal Writing Surface
12-565-170N Round 1.8 mL Internal Writing Surface
12-565-162N Round (Self-standing) 3.6 mL Internal Writing Surface, A-R Base
12-565-172N Round 3.6 mL Internal Writing Surface
12-566-98 Round 3.6 mL Internal Writing Surface, No Cap
12-565-291 Round (Self-standing) 4.5 mL External Writing Surface, A-R Base
12-565-161N Conical (Self-standing) 4.5 mL Internal Writing Surface, A-R Base
12-565-173N Round 4.5 mL Internal Writing Surface
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 12-565-299 Supplier Thermo Scientific™ Supplier No. 347643
Add to Cart
Add to Cart

Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.

Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.

  • Non-cytotoxic, non-mutatgenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • External or internally threaded options available
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Diameter (English) Outer 0.492 in.
Autoclavable Not Autoclavable
Capacity (Metric) 4.5 mL
Bottom Shape Round (Self-standing)
Material Polypropylene
Thread Style External
Length (English) 3.583 in.
Length (Metric) 91 mm
Packaging 300
Style No Writing Surface, A-R Base
RNase Free Yes
Vapor LN2 (to -150°C) Yes
Product Line Nunc
Sterility Sterile
Anti-Rotation Yes
Packaging Quantity 500 Pk.
Closure Material Polyethylene
Capacity (English) 0.1521 oz.
Dnase Free Yes
Closure Type Screw Cap
Printed No
Diameter (Metric) 12.5 mm
Product Type Cryogenic Storage Tube
Show More Show Less
Will the heat required to properly seal Nunc CryoFlex Tube Wrap adversely affect my cells?

Nunc Biobanking and Cell Culture Cryogenic Tubes are normally placed in a 4 degrees C ice bath during cell inoculation in the cryogenic tube. The heat required to shrink the polyethylene Nunc CryoFlex Tube Wrap will not raise the internal temperature enough to damage the cells. At this stage, cryoprotective agents are added.

What protocol should I follow for thawing cells, if my Nunc Biobanking and Cell Culture Cryogenic Tubes were previously immersed in liquid nitrogen without Nunc CryoFlex Tube Wrap?

We recommend the following protocol for thawing cells immersed in liquid nitrogen without Nunc CryoFlex Tube Wrap:
- Wear appropriate protective clothing including gloves, face guard, etc., during all transfer processes.
- The Nunc Biobanking and Cell Culture Cryogenic Tubes should be stored at least 24 hr in the gas phase of the liquid nitrogen freezer. This should allow trapped liquid nitrogen to evaporate and escape slowly, thereby reducing the possible pressure in the cryogenic tube.
- After at least 24 hr storage in the gas phase, the cryogenic tube may be put into a water bath and covered with a towel.
- After thawing, the contents of the cryogenic tube can be transferred.

Can I immerse Nunc Biobanking and Cell Culture Cryogenic Tubes in liquid nitrogen (liquid phase) for storage?

Of the Nunc Biobanking and Cell Culture Cryogenic Tubes, we recommend the internally threaded design, incorporating a silicon gasket, for liquid nitrogen immersion. At ultra low temperatures, any sealing material becomes inflexible. A theoretical risk of leakage is present when any mechanical closure is used. To facilitate leakproof storage, mechanically closed containers must be hermetically sealed in a protective envelope.

The Nunc cryo products includes Nunc CryoFlex Tube Wrap, a heat sealable polyethylene tubing designed for cryogenic tube encasement. The use of cryogenic tubes for freezing in liquid nitrogen (liquid phase) storage without CryoFlex tubing poses a safety hazard.
WARNING: Do not use Nunc Biobanking and Cell Culture Cryogenic Tubes in liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex Tube Wrap (Cat. No. 343958). Improper use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up, resulting in possible explosion or biohazard release. Use appropriate safety procedures as outlined in the Nalge Nunc International Cryopreservation Manual when handling and disposing of the vials.

What is the difference between your Nunc Biobanking and Cell Culture Cryogenic Tubes with internal threads and external threads?

Internally threaded Nunc Biobanking and Cell Culture Tubes are designed for vapor phase liquid nitrogen freezing. The cap incorporates a silicon gasket to enhance sealing properties.
Externally threaded Nunc Biobanking and Cell Culture Tubes are designed for mechanical freezing. The external screw cap reduces risk of contamination due to handling.

Can I autoclave Nunc Biobanking and Cell Culture Cryogenic Tubes?

We do not recommend autoclaving Nunc Biobanking and Cell Culture Tubes, even though they are made of polypropylene with polyethylene caps and stoppers.

Should I choose externally or internally threaded cryostorage vials?

Both types are designed and tested for secure cryogenic storage, so it’s largely a matter of personal preference. In an externally threaded vial (in which the cap covers the entire top portion of the vial itself), the underside of the cap is protected when unscrewed), which can minimize the potential for cross-contamination. In an internally threaded vial (in which the vial threads are inside the neck area and the cap inserts like a plug), the cap is the same diameter as the vial itself which can be helpful in certain types of dense storage applications.

Can I use your Nunc cryotubes or Nalgene cryotubes in liquid nitrogen?

Liquid nitrogen has very low surface tension, so it can flow into any of our cryostorage vials even if they are tightly capped. This creates significant potential for cross-contamination in storage as well as risk of injury from explosion when the vials are removed from storage and the liquid nitrogen boils off. The only option to mitigate this is Nunc Cryoflex tubing (e.g., Cat. No. 343958), which results in a heat-sealed barrier layer around each vial. Alternatively, storage in the vapor phase of liquid nitrogen is acceptable unless otherwise noted.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.