Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories
GREENER_CHOICE
Catalog No. 125651191
Change view
Click to view available options
Product Type:
Adapter Closure
Airvent Filter
Cover Cap
FEP Connector
Filter Assembly, 200 cm⊃2
Filter Assembly, 37 mm
PC Connector
Tubing Set, Female MPC
Tubing Set, Male MPC
Vent Cap
For Use With (Equipment):
Cell Factory Systems (All)
EasyFill Cell Factory Systems
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
12-565-1191 FEP Connector Cell Factory Systems (All)
12-566-956 PC Connector Cell Factory Systems (All)
12-566-340 Vent Cap EasyFill Cell Factory Systems
12-565-1193 Cover Cap Cell Factory Systems (All)
12-566-132 Airvent Filter Cell Factory Systems (All)
12-566-346 Filter Assembly, 200 cm⊃2 EasyFill Cell Factory Systems
12-566-342 Filter Assembly, 37 mm EasyFill Cell Factory Systems
12-567-219 Tubing Set, Female MPC Cell Factory Systems (All)
12-567-220 Tubing Set, Male MPC Cell Factory Systems (All)
12-566-343 Adapter Closure EasyFill Cell Factory Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 12-565-1191 Supplier Thermo Scientific™ Supplier No. 173250
Add to Cart
Add to Cart
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Connections made easy
  • Plug a media bag or tubing set into any cell factory
Save time and reduce costs
  • Cleaning processes and sterility validations are minimized
  • Most accessories are gamma irradiated and individually wrapped (Note: Connector Cat. Nos- 140099 and 173250 are not available gamma irradiated)
Prevent buildup of back pressure during filling application
  • Venting options provide additional air venting when filling and harvesting by gravity or with a peristaltic pump
TRUSTED_SUSTAINABILITY
Specifications
Description FEP Connector (Autoclavable)
Material FEP
For Use With (Equipment) Cell Factory Systems (All)
No. per Case 4
No. per Pack 1
Product Type FEP Connector
Autoclavable Autoclavable
Sterility Non-sterile
Bottom Shape Flat
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.