Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories
GREENER_CHOICE
Catalog No. 12566340
Change view
Click to view available options
Product Type:
Adapter Closure
Airvent Filter
Cover Cap
FEP Connector
Filter Assembly, 200 cm⊃2
Filter Assembly, 37 mm
PC Connector
Tubing Set, Female MPC
Tubing Set, Male MPC
Vent Cap
For Use With (Equipment):
Cell Factory Systems (All)
EasyFill Cell Factory Systems
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
12-566-340 Vent Cap EasyFill Cell Factory Systems
12-566-956 PC Connector Cell Factory Systems (All)
12-565-1193 Cover Cap Cell Factory Systems (All)
12-566-132 Airvent Filter Cell Factory Systems (All)
12-566-346 Filter Assembly, 200 cm⊃2 EasyFill Cell Factory Systems
12-566-342 Filter Assembly, 37 mm EasyFill Cell Factory Systems
12-567-219 Tubing Set, Female MPC Cell Factory Systems (All)
12-567-220 Tubing Set, Male MPC Cell Factory Systems (All)
12-565-1191 FEP Connector Cell Factory Systems (All)
12-566-343 Adapter Closure EasyFill Cell Factory Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 12-566-340 Supplier Thermo Scientific™ Supplier No. 146008
Add to Cart
Add to Cart
This item is not returnable. View return policy
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Connections Made Easy:
  • Plug a media bag or tubing set to any Cell Factory system for enhanced aseptic connectivity for filling and harvesting
Additional Features:
  • Save time and reduce costs
  • Minimize cleaning processes such as autoclaving and associated validations
  • Prevent build up of back pressure during filling application
  • Get additional air venting with the venting options
TRUSTED_SUSTAINABILITY
Specifications
Description Easy Fill Replacement Vent Cap
Material HDPE
For Use With (Equipment) EasyFill Cell Factory Systems
No. per Case 10
No. per Pack 10
Product Type Vent Cap
Green Features Fewer resources used and less waste, Sustainable packaging
Bottom Shape Flat
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.