Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Cell Factory™ System
GREENER_CHOICE

Catalog No. 1256538
Change view
Click to view available options
Tray Layers:
1
2
4
10
40
Culture Area:
1264 cm²
25,280 cm²
2528 cm²
632 cm²
6320 cm²
No. per Case:
10/Cs.
2/Cs.
6/Cs.
8/Cs.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tray Layers Culture Area No. per Case
12-565-38 2 1264 cm² 6/Cs.
12-565-289 40 25,280 cm² 2/Cs.
12-566-90 4 2528 cm² 10/Cs.
12-565-39 10 6320 cm² 2/Cs.
12-565-37 1 632 cm² 8/Cs.
12-565-40 10 6320 cm² 6/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12-565-38 Supplier Thermo Scientific™ Supplier No. 167695
Add to Cart
Add to Cart
This item is not returnable. View return policy

Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.

Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.

  • Ideal for adherent cells
  • Boost production efficiency with more surface area in a small footprint
  • Reduce contamination risks with a closed system
Warranty: 90 days
TRUSTED_SUSTAINABILITY

Specifications

Description Original Cell Factory
Material Polystyrene
No. per Case 6/Cs.
No. per Pack 1 Ea.
Product Type Cell Factory System
Surface Coating Nunclon™ Delta
Green Features ISCC PLUS
Closure Type Adapter Cap and Port Cover Cap
Culture Area 1264 cm²
Surface Treatment Nunclon Delta
Tray Layers 2
Show More Show Less
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

What volume of medium should I use for Nunc Cell Factory Systems?

We recommend a 200 mL volume of medium per tray (632 cm^2).
Therefore the minimum volumes we recommend are as follows:
Cat. No.; No. per Pack/No. per Case; No. of Trays; Minimum Volume (mL)
- 165250; 1/8; 1-Tray; 200 mL
- 167695; 1/5; 2-Tray; 400 mL
- 164327; 1/2; 10-Tray; 2000 mL
- 170009; 1/6; 10-Tray; 2000 mL
- 139446; 1/2; 40-Tray; 8000 mL

What are the dimensions and culture areas for each tray version of your Nunc Cell Factory Systems?

Please review the following information regarding the dimension and culture areas for each tray version of the Nunc Cell Factory Systems:
Cat. No.; No. per Pack/No. per Case; No. of Tray Layers; Dimensions L x W x H (mm); Culture Area (cm^2)
- 165250; 1/8; 1-Tray; 335 mm x 205 mm x 37 mm; 632 cm^2
- 167695; 1/6; 2-Tray; 335 mm x 205 mm x 52 mm; 1264 cm^2
- 164327; 1/2; 10-Tray; 335 mm x 205 mm x 190 mm; 6320 cm^2
- 170009; 1/6; 10-Tray; 335 mm x 205 mm x 190 mm; 6320 cm^2
- 139446; 1/2; 40-Tray; 335 mm x 205 mm x 700 mm; 25284 cm^2

Can I view cells in a 10-tray Nunc Cell Factory System under a microscope?

Due to the height of the 10-tray and 40-tray Nunc Cell Factory Systems, cells cannot be viewed with common microscopes. Typically cells are seeded in a 1-tray or 2-tray cell factory system as a control. Cells grown in each of these products can be viewed under a microscope.
Alternatively, inverted stereo microscopes, with a powerful light source on the view side, and adjusted for the height of the cell factory system have been used to view the first few layers.

What are your Nunc Cell Factory Systems?

Our Nunc Cell Factory Systems are designed for large scale cell culture and production of biomaterials such as vaccines, monoclonal antibodies, etc. They provide large growth surface in limited space areas and have the Nunclon Delta surface coating. Some benefits are the low risk of contamination and the easy to handle multi-layer stacks. Nunc Cell Factory Systems are available with 1-, 2-, 10-, and 40-tray versions.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.