Learn More
Thermo Scientific™ Nunc™ 384-Well Polypropylene DeepWell™ Storage Plate
GSA_VAGREENER_CHOICE
Catalog No. 12565229 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
60/Cs.
Catalog No. Quantity
12-565-229 60/Cs.
1 options
Catalog No. 12-565-229 Supplier Thermo Scientific™ Supplier No. 269390
  • $1,172.00
    $802.65 / Case of 60
    Save $369.35
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Simplify compound library storage with the Thermo Scientific™ Nunc™ DeepWell™ 384-Well Plate.
Simplify compound library storage with the Thermo Scientific™ Nunc™ DeepWell™ 384-Well Plate. This deepwell polypropylene plate provides larger working volumes and minimizes wicking through a special rounded square well design. Polypropylene is ideal for storage plates because it has a lower binding capacity, allowing for complete sample recovery.
Features:
  • Raised rims around each well are ideal for heat or adhesive sealing
  • Rounded wells are for optimized mixing
  • Compatible with robotics, automation and liquid handling systems
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Description 384-well Polypropylene Sample Processing & Storage Plates
Array 16 x 24
Autoclavable Autoclavable
Packaging Individually Wrapped
Binding Type Low binding PP
Product Line Nunc
Color Natural
Product Type 384-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 240 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Square, Round Bottom (U-well)
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Green Features ACT Ecolabel (Silver)
No. per Case 60
No. per Pack 1
Quantity 60/Cs.
Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
RNase Free Yes
Fit for Purpose PCR Ready
Plate Size ANSI/ SLAS Standard
Show More Show Less
For Research Use Only. Not for use in diagnostic procedures.