Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Microplate Lids
GREENER_CHOICE

Catalog No. 12565604
Change view
Click to view available options
Plate Compatibility:
384-well Plate
96-well Plate
All Microplates
Sterility:
Non-sterile
Sterile
Condensation Rings:
No
Yes
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Compatibility Sterility Condensation Rings
12-565-604 384-well Plate Non-sterile No
12-565-476 All Microplates Sterile No
12-565-478 All Microplates Non-sterile No
12-565-462 All Microplates Non-sterile No
12-565-748 All Microplates Non-sterile Yes
12-565-749 All Microplates Sterile Yes
12-565-69 96-well Plate Non-sterile Yes
12-565-70 96-well Plate Sterile Yes
12-565-603 384-well Plate Sterile No
12-565-477 96-well Plate Sterile Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 12-565-604 Supplier Thermo Scientific™ Supplier No. 264612
Add to Cart
Add to Cart

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Dimensions (L x W x H) 5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
Material Polystyrene
Sterility Non-sterile
No. of Wells 384
Height (Metric) 7.9 mm
Length (English) 5 in.
Length (Metric) 127 mm
Condensation Rings No
Height (English) 0.3 in.
No. per Case 180
No. per Pack 20
Dimensions (L x W) 5 x 3.35 in. (127 x 85 mm)
Evaporation Barrier Yes
Width (English) 3.35 in.
Width (Metric) 85 mm
Product Type Plate Lid
Product Line Nunc
Adhesive None
Temperature Range (Metric) -20°C to +80°C
Well Shape All well shapes
Plate Compatibility 384-well Plate
Cut-off Corners Long Side
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.