Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ PETG Roller Bottles
GREENER_CHOICE

Catalog No. 12565532
Change view
Click to view available options
Cell Growth Area:
1050 cm2
1700 cm2
1800 cm2
2100 cm2
4200 cm2
Bottle Surface:
Pleated
Smooth
Packaging:
20/case
22/case
5/sleeve, 20/case
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Growth Area Bottle Surface Packaging
12-565-532 4200 cm2 Pleated 22/case
12-565-526 1050 cm2 Smooth 5/sleeve, 20/case
12-565-527 1050 cm2 Smooth 20/case
12-575-120 1050 cm2 Smooth 5/sleeve, 20/case
12-565-528 1700 cm2 Pleated 20/case
12-565-529 1800 cm2 Smooth 22/case
12-565-531 2100 cm2 Pleated 20/case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 12-565-532 Supplier Thermo Scientific™ Supplier No. 426022
Add to Cart
Add to Cart
This item is not returnable. View return policy

Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.

Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.

Key Features:
  • Molded of durable PETG
  • Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
  • Provides solutions for all scales with wide range of surface areas from 1050cm2 to 4200cm2
  • Reduces wrist strain and increases productivity with quick-action ergonomic closure
  • Can be traced by lot number on every bottle
Recommended for:
  • Cell culture expansion
  • Vaccine production
  • Antibody production
TRUSTED_SUSTAINABILITY

Specifications

Bottle Surface Pleated
Color Clear
Description 4260-22
Material PETG
No. per Case 22
No. per Pack 22
Packaging 22/case
Cell Growth Area 4200 cm2
Closure Included Yes
Graduated Yes
Shape Round
Product Type Roller Bottle
Closure Type Quick-Action Ergonomic
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.