Learn More
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles
Catalog No. 12565683
Change view
Click to view available options
Cell Growth Area:
1450 cm2
850 cm2
Bottle Surface:
Pleated, no indent
Smooth, no indent
Closure Type:
Easy on/off, Non-vented Cap
Easy on/off, Vented Cap
Packaging:
2/Pk., 20/Cs.
20/Pk., 20/Cs.
20/Pk., 20/Cs. (Double-bagged)
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Growth Area Bottle Surface Closure Type Packaging
12-565-683 1450 cm2 Pleated, no indent Easy on/off, Vented Cap 20/Pk., 20/Cs. (Double-bagged)
12-565-685 850 cm2 Smooth, no indent Easy on/off, Vented Cap 2/Pk., 20/Cs.
12-565-687 850 cm2 Smooth, no indent Easy on/off, Non-vented Cap 20/Pk., 20/Cs.
12-565-688 850 cm2 Smooth, no indent Easy on/off, Non-vented Cap 20/Pk., 20/Cs. (Double-bagged)
12-565-686 850 cm2 Smooth, no indent Easy on/off, Non-vented Cap 2/Pk., 20/Cs.
12-565-684 1450 cm2 Pleated, no indent Easy on/off, Non-vented Cap 20/Pk., 20/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12-565-683 Supplier Thermo Scientific™ Supplier No. 141744
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.
Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6.
Features:
  • Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6
  • One-piece seamless design reduces possibility of leaking through a seam.
  • Shallow and deep indents allow for excellent manual and automated handling.
  • Proprietary manufacturing process allows for greater impact resistance
  • Vertical expanded surfaces version facilitates emptying and reduces retention of product
Recommended for:
  • Cell culture expansion
  • Vaccine production
  • Antibody production
TRUSTED_SUSTAINABILITY
Specifications
Bottle Surface Pleated, no indent
Material Polystyrene
No. per Case 20
No. per Pack 20
Packaging 20/Pk., 20/Cs. (Double-bagged)
Cell Growth Area 1450 cm2
Closure Included Yes
Product Type Roller Bottle
Closure Type Easy on/off, Vented Cap
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.