Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP214 Rabbit anti-Human, Mouse, Polyclonal, liquid, OriGene
SDP

Catalog No. 501684823
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4823 Supplier OriGene Supplier No. TA890139
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcineurin is a widely distributed Ser/Thr, calcium and calmodulin-dependent protein phosphatase. It mediates the immunosuppressive actions of drugs such as FK506 and cyclosporin, and has also been implicated in a number of calcium-sensitive pathways in the nervous system. Calcineurin has been found to be associated with other proteins, such as calmodulin, FKBP12, the ryanodine receptor, inositol 1,4,5-trisphosphate, and a recently identified protein, calcineurin inhibitor (cain). Cain has been shown to bind calcineurin through its C-terminal tail. Recent studies have shown cain to bind to both amphiphysin 1 and calcineurin simultaneously and therefore acts as a physiological inhibitor of calcineurin. The binding of cain to amphiphysin 1 does not affect the interaction of amphiphysin 1 with other endocytic proteins. Cain, like calcineurin, is highly expressed in neurons and other tissues.

Specifications

Antigen NUP214
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NUP214
Gene Accession No. P35658, Q80U93
Gene Alias CAIN, CAN
Gene Symbols NUP214
Host Species Rabbit
Immunogen Recombit protein of human NUP214
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227720, 8021
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.