Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566007 Supplier Invitrogen™ Supplier No. PA566007
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YPQNKRPVHMD RACLFRWSTI SENLFATTGY PGKMASQFQI HHLGHPQPIL MGSVAVGSGL SWHRTLPLCV IGGDHKLLFW VTEV Highest antigen sequence identity to the following orthologs - mouse 88%, rat 86%.

Component of the Nup107-160 subcomplex of the nuclear pore complex (NPC). The Nup107-160 subcomplex is required for the assembly of a functional NPC. The Nup107-160 subcomplex is also required for normal kinetochore microtubule attachment, mitotic progression and chromosome segregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP37
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUP37
Gene Accession No. Q8NFH4
Gene Alias 2410003L22Rik; 2810039M17Rik; nucleoporin 37; nucleoporin 37kDa; nucleoporin Nup37; nucleoporin Nup37-like protein; nup107-160 subcomplex subunit Nup37; NUP37; p37
Gene Symbols NUP37
Host Species Rabbit
Immunogen Recombinant Human NUP37. Recombinant protein control fragment (Product #RP-105038).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79023
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.