Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555572 Supplier Invitrogen™ Supplier No. PA555572
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HAVTFLRTPE ILTVNSIGQL KIWDFRQQGN EPSQILSLTG DRVPLHCVDR HPNQQHVVAT GGQDGMLSIW DV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Component of the Nup107-160 subcomplex of the nuclear pore complex (NPC). The Nup107-160 subcomplex is required for the assembly of a functional NPC. The Nup107-160 subcomplex is also required for normal kinetochore microtubule attachment, mitotic progression and chromosome segregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP43
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUP43
Gene Accession No. P59235, Q8NFH3
Gene Alias 2610016K01Rik; 2610529I12Rik; AA409950; bA350J20.1; nucleoporin 43; nucleoporin 43kDa; nucleoporin Nup43; Nup107-160 subcomplex subunit Nup43; Nup43; p42
Gene Symbols NUP43
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUP43. Recombinant protein control fragment (Product #RP-94722).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348995, 683983, 69912
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.