Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP62 Monoclonal Antibody (RL31)
GREENER_CHOICE

Catalog No. MA1073 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1073 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1073 Supplier Invitrogen™ Supplier No. MA1073
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

MA1-073 detects the nucleoporin p62 O-linked glycoprotein in the p62 complex from rat, human and non-human primate tissues. MA1-073 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects a single ~62 kDa protein representing nucleoporin p62 in rat liver nuclear envelope fractions. Immunofluorescence staining of p62 in rat liver with MA1-073 results in staining of both the nucleoplasmic and cytoplasmic surfaces of the nuclear pore complex (NPC). The MA1-073 immunogen is O-linked glycoprotein-enriched fraction from rat liver nuclear envelopes raised in Armenian hamsters.

SDS3 is a subunit of the histone deacetylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP62
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone RL31
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.2-7.4
Gene Nup62
Gene Accession No. P17955, P37198, Q63850
Gene Alias 62 kDa nucleoporin; AA589433; AI426861; AI790512; AU045898; D7Ertd649e; DKFZp547L134; FLJ20822; FLJ43869; IBSN; MGC841; Np62; nuclear pore complex 1; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin Nup62; nucleoporin p62; Nup62; Nupc1; p62; SNDI
Gene Symbols Nup62
Host Species Armenian Hamster
Immunogen O-linked glycoprotein-enriched fraction from rat liver nuclear envelopes raised in Armenian hamsters.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18226, 23636, 484370, 65274
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.