Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP88 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103980 Supplier Invitrogen™ Supplier No. PA5103980
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NUP88.

The nuclear pore complex (NPC) mediates bidirectional macromolecular traffic between the nucleus and cytoplasm in eukaryotic cells and is comprised of more than 100 different subunits. Many of the subunits belong to a family called nucleoporins (Nups), which are characterized by the presence of O-linked-N-acetylglucosamine moieties and a distinctive pentapeptide repeat (XFXFG). Nup88 (nucleoporin 88 kDa) is a 741 amino acid protein that localizes to the nucleus and functions as an essential component of the nuclear pore complex. Expressed ubiquitously, Nup88 is subject to phosphorylation by ATM or ATR and is upregulated in malignant neoplasms and precancerous dysplasias, suggesting a role in tumorigenesis. The gene encoding Nup88 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1, 200 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP88
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NUP88
Gene Accession No. Q8CEC0, Q99567
Gene Alias 88 kDa nuclear pore complex protein; 88 kDa nucleoporin; karyoporin; nuclear pore complex protein 88; nuclear pore complex protein Nup88; nucleoporin 88; nucleoporin 88kDa; nucleoporin Nup84; Nucleoporin Nup88; Nup84; NUP88; Prei2; preimplantation protein 2
Gene Symbols NUP88
Host Species Rabbit
Immunogen A synthesized peptide derived from human NUP88(Accession Q99567), corresponding to amino acid residues D436-T486.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19069, 4927
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.