Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUP93 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553674 Supplier Invitrogen™ Supplier No. PA553674
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VERNLQEIQQ AGERLRSRTL TRTSQETADV KASVLLGSRG LDISHISQRL ESLSAATTFE PLEPVKDTDI QGFLKNEKDN ALLSAIEESR KRTFGMAEEY HRESMLVEWE QVKQRI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. This gene encodes a nucleoporin protein that localizes both to the basket of the pore and to the nuclear entry of the central gated channel of the pore. The encoded protein is a target of caspase cysteine proteases that play a central role in programmed cell death by apoptosis. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP93
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nup93
Gene Accession No. Q66HC5, Q8BJ71, Q8N1F7
Gene Alias 2410008G02Rik; 93 kDa nucleoporin; Allele: hi4; CBP-interacting protein 4; Cip4; dead eye; dead eye protein; dye; Kiaa0095; NIC96; NPHS12; nuclear pore complex protein Nup93; nucleoporin 93; nucleoporin 93kDa; nucleoporin Nup93; nup93
Gene Symbols Nup93
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUP93. Recombinant protein control fragment (Product #RP-91837).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291874, 71805, 9688
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.