Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NuPAGE™ WedgeWell™ Midi Tris-Acetate Welcome Pack, 3 to 8%

Catalog No. STM5007
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
20-well
26-well
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
STM5007 26-well
STM5006 20-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. STM5007 Supplier Invitrogen™ Supplier No. STM5007
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The NuPAGE Tris-Acetate Midi Welcome Pack, Wedgewell format provides all the necessary NuPAGE gels, buffers, and reagents needed to begin using the SureLock Tandem Midi Gel Tank.

The NuPAGE Tris-Acetate Midi Wedgewell Welcome Pack provides all the necessary NuPAGE gels, buffers, and reagents needed to begin using the SureLock Tandem Midi Gel Tank.

Welcome Pack contents

  • SureLock Tandem Midi Gel Tank (STM1001)
  • NuPAGE Tris-Acetate WedgeWell Midi Gels (2 boxes, 20 gels)
  • NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • HiMark Pre-stained Protein Standard (LC5699)

About the SureLock Tandem Midi Gel Tank

The SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a setup time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.

About NuPAGE Tris-Acetate Midi gels

NuPAGE Tris-Acetate Midi protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate Midi gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine midi SDS-PAGE gels.

Transfer and detection

For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the SureLock Tandem Midi Blot Module (STM2001). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Convenient to use

NuPAGE WedgeWell-format Tris-Acetate gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) SureLock Tandem Midi Gel Tank
Gel Size Midi
Product Line NuPAGE
Gel Percentage 3 to 8%
Gel Type Tris-Acetate
No. of Wells 26-well
Quantity 1 Welcome Pk.
Separation Range 40 to 500 kDa
Shipping Condition Approved for shipment at room temperature and wet ice
Thickness (Metric) 1.0 mm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.