Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NuPAGE™ WedgeWell™ Tris-Acetate Welcome Pack and Mini Blot Modules, 3 to 8%

Catalog No. TA03810G
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
TA03810G 10-well
TA03812H 12-well
TA03815I 15-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. TA03810G Supplier Invitrogen™ Supplier No. TA03810G
Get Up to 55% Off Protein Gel Welcome Packs
View all promotions
Add to Cart
Add to Cart

The NuPAGE Tris-Acetate Welcome Pack and Mini Blot Modules provide all the necessary NuPAGE gels and buffers needed to begin using the Mini Gel Tank plus two Mini Blot modules for wet transfer.

The NuPAGE Tris-Acetate WedgeWell Welcome Pack and Mini Blot Modules provide all the necessary NuPAGE gels and buffers needed to begin using the Mini Gel Tank plus two Mini Blot modules for wet transfer.

Welcome Pack contents

  • Mini Gel Tank (A25977)
  • NuPAGE Tris-Acetate WedgeWell mini gels (2 boxes, 20 gels)
  • NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • HiMark Pre-stained Protein Standard (LC5699)
  • 2 x Mini Blot Module (B1000)

About the Mini Gel Tank

The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use.

About NuPAGE Tris-Acetate gels

NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.

Transfer

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (Cat. No. NP0006) for traditional wet transfer using the Mini Blot Module (B1000).

About the Mini Blot Module

The Mini Blot Module is a wet transfer device used exclusively with the Mini Gel Tank. The tank accommodates one blot module per chamber or two blot modules total with the side-by-side layout. The Mini Blot Module is compatible all Invitrogen mini gels (blot size: 8.5 x 8 cm). The blot module accommodates one mini gel per wet transfer. The universal connection and molded gasket make the module easy to use, while the inner core of the module allows for use of less methanol-based transfer buffer per wet transfer (250 mL per gel). At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes in 60 minutes.

Convenient to use

NuPAGE Tris-Acetate WedgeWell-format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) XCell SureLock Mini-Cell
Gel Size Mini
Product Line NuPAGE
Gel Percentage 3 to 8%
Gel Type Tris-Acetate
No. of Wells 10-well
Quantity 1 Welcome Pk. kit
Separation Range 40 to 500 kDa
Shipping Condition Approved for shipment at room temperature and wet ice
Thickness (Metric) 1.0 mm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.