Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NuPAGE™ WedgeWell™ Tris-Acetate XCell SureLock™ Mini-Cell Welcome Pack, 3 to 8%

Catalog No. TA03812E
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
TA03812E 12-well
TA03810D 10-well
TA03815F 15-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. TA03812E Supplier Invitrogen™ Supplier No. TA03812E
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The NuPAGE Tris-Acetate Welcome Pack, WedgeWell format provides all the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.

The NuPAGE Tris-Acetate WedgeWell Welcome Pack provides all the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.

Welcome Pack contents

  • XCell SureLock Mini-Cell (EI0001)
  • NuPAGE Tris-Acetate mini WedgeWell gels (2 boxes, 20 gels)
  • NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • HiMark Pre-stained Protein Standard (LC5699)

About the XCell SureLock Mini-Cell

The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and SureCast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Tris-Acetate gels

NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer buffer (NP0006) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Convenient to use

NuPAGE Tris-Acetate WedgeWell-format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) XCell SureLock Mini-Cell
Gel Size Mini
Product Line NuPAGE
Gel Percentage 3 to 8%
Gel Type Tris-Acetate
No. of Wells 12-well
Quantity 1 Welcome Pk. kit
Separation Range 40 to 500 kDa
Shipping Condition Approved for shipment at room temperature and wet ice
Thickness (Metric) 1.0 mm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.