Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NusA Monoclonal Antibody (A6-E7)
GREENER_CHOICE

Catalog No. PIMA542570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542570 Supplier Invitrogen™ Supplier No. MA542570
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Positive Control: E.coli Subcellular Location: Cytoplasm.

NusA is a key component in both prevention and enhancement of transcriptional termination. It is important in both Rho-dependent and intrinsic termination, as well as in lambda and other phage antitermination systems. The gene was first identified by isolation of the nusAl mutation, which restricts bacteriophage l growth by preventing the antitermination activity of the l N protein. NusA is involved in transcriptional antitermination in the cell. It has been shown to specifically aid in read-through of the RNA polymerase genes rpoB and rpoC, as well as in successful synthesis of the ribosomal RNA genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NusA
Applications ELISA, Western Blot
Classification Monoclonal
Clone A6-E7
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene nusA
Gene Accession No. 0
Gene Symbols nusA
Host Species Mouse
Immunogen Recombinant full length protein of Escherichia coli NusA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 947682
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.