Learn More
Ampco Safety Tools Nutdriver (Imperial)
SDP
Catalog No. 19174186 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
1/2 in.
1/4 in.
1/8 in.
11/16 in.
11/32 in.
13/16 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/32 in.
7/8 in.
9/16 in.
9/32 in.
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-174-186 9/16 in.
19-174-152 1/2 in.
19-174-153 1/4 in.
19-174-154 1/8 in.
19-174-156 11/16 in.
19-174-157 11/32 in.
19-174-160 13/16 in.
19-174-168 3/16 in.
19-174-169 3/4 in.
19-174-170 3/8 in.
19-174-174 5/16 in.
19-174-175 5/32 in.
19-174-176 5/8 in.
19-174-181 7/16 in.
19-174-182 7/32 in.
19-174-183 7/8 in.
19-174-187 9/32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-186 Supplier Ampco Safety Tools Supplier No. ND9/16
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Nutdriver performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Length (English) 9/16 in.
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.