Learn More
Ampco Safety Tools Nutdriver (Metric)
SDP
Catalog No. 19174185 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
3.5 mm
4 mm
4.5 mm
5 mm
5.5 mm
6 mm
6.5 mm
7 mm
7.5 mm
8 mm
9 mm
10 mm
11 mm
12 mm
13 mm
14 mm
15 mm
16 mm
17 mm
18 mm
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-174-185 8 mm
19-174-155 10 mm
19-174-158 11 mm
19-174-159 12 mm
19-174-161 13 mm
19-174-162 14 mm
19-174-163 15 mm
19-174-164 16 mm
19-174-165 17 mm
19-174-166 18 mm
19-174-167 3.5 mm
19-174-171 4.5 mm
19-174-172 4 mm
19-174-173 5.5 mm
19-174-177 5 mm
19-174-178 6.5 mm
19-174-179 6 mm
19-174-180 7.5 mm
19-174-184 7 mm
19-174-188 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-185 Supplier Ampco Safety Tools Supplier No. ND8MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Nutdriver performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Length (Metric) 8 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.