Learn More
Enzo Life Sciences Nutlin-3 (1mg). CAS: 548472-68-0
SDP
Supplier:  Enzo Life Sciences ALX430128M001
Encompass_Preferred
(+/-) -Nutlin-3 is an inhibitor of p53/MDM2 interaction (IC50=0.09µM). Alternative name: (+/-)-4-[4,5-bis-(4-Chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydroimidazole-1-carbonyl]-piperazin-2-one. Purity: ≥98% (HPLC (UV)). Solubility: Soluble in methanol, 100% ethanol, DMSO, dimethyl formamide or acetonitrile; sparingly soluble in a 1:10 solution of ethanol:PBS, pH 7.2. Long Term Storage: -20°C.
Catalog No. 50-200-9216
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.