Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nutlin-3, Thermo Scientific™

Catalog No. AC469201000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469201000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC469201000 Supplier Thermo Scientific Chemicals Supplier No. 469201000
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 548472-68-0
Molecular Formula C30 H30 Cl2 N4 O4

Specifications

Quantity 100 mg
Formula Weight 581.5
Physical Form Crystalline Powder
Color White to Light-yellow
Purity ≥97.5%
Chemical Name or Material Nutlin-3
Hazard Category Exclamation mark
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.