Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Nutrient Broth

Catalog No. MP091007917 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP091007917 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091007917 Supplier MP Biomedicals™ Supplier No. 091007917
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nutrient Broth, Tan Powder

  • Nutrient Broth is used in many laboratory procedures as is, or with added indicators, carbohydrates, organic liquids, salts, etc.
  • This medium is used in accordance with official recommended procedures for the bacteriological analyses of water, milk and dairy products, in foods of important sanitation, tests for sensitivity and resistance and as a base to prepare media supplemented with other nutrients.
  • It can be used for enumeration, isolation and enrichment as well as providing a high grade base for the preparation of special media.
  • With the addition of blood, ascites fluid or serum, they can also be used to cultivate streptococci, pneumococci and erysipelas species etc.
  • Nutrient Broth is a general purpose medium used for cultivating a broad variety of fastidious and non-fastidious microorganisms with non-exacting nutritional requirements.
  • Peptone and yeast extract provide nitrogenous compounds, vitamin B complex, amino acids and other essential growth nutrients.
  • Sodium chloride is used to maintain the osmotic balance and glucose is the fermentable carbohydrate source of the media.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature (15-30°C)
Form Tan Powder
Preparation Method Sterilize at 121°C (15 lbs. of pressure) for 15 minutes.
Product Type Microbiology Media
Formulation Nutrient Broth is a liquid medium, produced according to the formula from APHA and AOAC. Approximate formula:gelatin Peptone: 5g/L; Beef Extract: 3g/L; Final pH: 6.9 +/- 0.1
For Use With (Application) Growth of fastidious and non-fastidious microorganisms
pH 6.7 - 7.1 (8g/L)
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.