Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nutrient Broth (Dehydrated)

Catalog No. CM0001R
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM0001R 2.5 kg
OXCM0001B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. CM0001R Supplier Thermo Scientific™ Supplier No. CM0001R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cultivate fastidious pathogens and other microorganisms with this nutritious medium.

Cultivate fastidious pathogens and other microorganisms with nutritious Thermo Scientific™ Oxoid™ Nutrient Broth (Dehydrated). This general purpose medium contains beef extracts combined with peptone, glucose, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.

A general purpose fluid medium for the cultivation of microorganisms not exacting in their nutritional requirements.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Dehydrated Culture Media
Quantity 2.5 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.