Learn More
MilliporeSigma™ Nuts for Hibar™/LiChroCART™ Columns
Catalog No. M1155450001 Shop All MilliporeSigma Products
Change view
Click to view available options
Includes:
PVDF double cones
Packaging Quantity:
4/Pk.
Catalog No. Includes Packaging Quantity
M1155450001 PVDF double cones 4/Pk.
1 options
Catalog No. M1155450001 Supplier MilliporeSigma™ Supplier No. 1155450001
Add to Cart
Add to Cart
Knurled nuts for capillary connection
The package contains knurled nuts.
TRUSTED_SUSTAINABILITY
Specifications
Type Nut
For Use With (Application) For Capillary Connection
Packaging Quantity 4/Pk.
Includes PVDF double cones
For Use With Hibar Columns/LiChroCART Cartridges