Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Invitrogen Collibri Stranded RNA Library Prep Kit

Catalog No. A39119196
Encompass_Preferred
Change view
Click to view available options
Product Type:
Library Cleanup Kit
RNA Single-indexes (Set 1-24)
RNA Single-indexes (Set 1-96)
RNA UD Indexes (Set 1-24)
RNA UD Indexes (Set 1-96)
Stranded RNA Library Prep Kit
rRNA Depletion Kit
Index Included:
No
Yes
Quantity:
24 Reactions
24 preps
96 Reactions
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Index Included Quantity
A39119196 RNA UD Indexes (Set 1-96) Yes 96 Reactions
A39100024 Library Cleanup Kit No 24 preps
A39114096 Library Cleanup Kit No 96 Reactions
A39115024 rRNA Depletion Kit No 24 Reactions
A39115096 rRNA Depletion Kit No 96 Reactions
A39117024 Stranded RNA Library Prep Kit No 24 Reactions
A39117096 Stranded RNA Library Prep Kit No 96 Reactions
A39118024 RNA Single-indexes (Set 1-24) Yes 24 Reactions
A39118096 RNA Single-indexes (Set 1-96) Yes 96 Reactions
A39119024 RNA UD Indexes (Set 1-24) Yes 24 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. A39119196 Supplier Thermo Scientific™ Supplier No. A39119196
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Invitrogen Collibri Stranded RNA Library Preparation Kit Modules are designed for a custom build of next-generation sequencing (NGS) workflow on Illumina systems. The modules originally support whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.

The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for the robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The modules bring together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality sequencing-ready libraries.

The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for the robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The modules bring together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality, sequencing-ready libraries.

This Collibri Stranded RNA Library Prep Kit Modules supports fast library preparation within 4.5 hours for mRNA or whole transcriptome libraries with 1–25 ng of mRNA-enriched or ribodepleted RNA sample input. Multiplexing of libraries can be carried out using up to 96 single-indexed or unique dual-indexed primers, enabling single-read or paired-end sequencing. The kit is suitable for various-quality RNA samples, including FFPE samples.

Visual cues of process integrity

For maximum convenience, visual cues of process integrity are included throughout the library preparation workflow. Monitor progress of library generation using visual feedback from dyes in critical reagents. The reaction mix changes color in response to the addition of critical components at each step to ensure that library preparation has the highest chance of success. The inert dyes do not interfere with enzymatic reactions and do not compromise sequencing results.

How it works

Full-length Illumina-compatible adaptors are introduced by PCR amplification using Platinum SuperFi DNA Polymerase to construct single-indexed or unique dual-indexed libraries compatible with single-read or paired-end sequencing. The kit contains 24 or 96 either single-indexed barcoded PCR primers (i7) pre-mixed with a universal i5 primer or unique dual-indexed PCR primers that enable multiplexing of up to 96 libraries. Optimized cleanup steps efficiently remove residual primers and adapter/primer dimers while preserving high library yields.

A related product, the Collibri Library Quantification Kit, is recommended for qPCR-based quantifications of libraries before proceeding to sequencing.

Applications

The Collibri Stranded RNA Library Prep Kit is recommended for:

  • Gene expression studies
  • Alternative splicing analysis
  • Non-coding RNA detection and discovery
  • Identification of alternative polyadenylation sites
  • Gene fusion detection
  • Detection of transcript isoforms
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C
Description Indexing
Format Kit
For Use With (Application) RNA Sequencing, Whole Transcriptome, mRNA Sequencing, Small RNA Sequencing
Product Line Collibri
Product Type RNA UD Indexes (Set 1-96)
Quantity 96 Reactions
Index Included Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.