Learn More
Selleck Chemical LLC NVP-ADW742 5mg 475488-23-4 GSK 552602A, ADW742
SDP
Supplier:  Selleck Chemical LLC S10885MG
Encompass_Preferred
NVP-ADW742 (GSK 552602A) is an IGF-1R inhibitor with IC50 of 0.17 ?M, >16-fold more potent against IGF-1R than InsR; little activity to HER2, PDGFR, VEGFR-2, Bcr-Abl and c-Kit. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0904
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.